Gene Transcripts

Gene Symbol
FLNC
Entrez Gene ID
2318
Gene Name
filamin C
Gene Aliases
ABP-280, ABP280A, ABPA, ABPL, FLN2, MFM5, MPD4
Chromosome Location
Chr. 7: 128830429 - 128859274 on Build GRCh38
UniGene ID
Hs.58414
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127487.1 NP_001120959.1 4 994
NM_001458.4 NP_001449.3 4 994
GenBank mRNA AB371585.1 - 4 1014
AF089841.1 - 4 834
AF146692.1 - 4 997
AJ012737.1 - 4 803

View Full Product Details