Gene Transcripts

Gene Symbol
Iqcg
Entrez Gene ID
69707
Gene Name
IQ motif containing G
Gene Aliases
2400003L07Rik, G1-374-12, esgd12d, repro1, stubby12d
Chromosome Location
Chr. 16: 33014270 - 33056341 on Build GRCm38
UniGene ID
Mm.227918
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178378.3 NP_848465.1 4, 5 559
GenBank mRNA AK010269.1 - 1, 2 110
AK146422.1 - 2, 3 297
BC049990.1 - 4, 5 543 6310844

View Full Product Details