Gene Transcripts
- Gene Symbol
- AMPH
- Entrez Gene ID
- 273
- Gene Name
- amphiphysin
- Gene Aliases
- AMPH1
- Chromosome Location
- Chr. 7: 38383694 - 38631567 on Build GRCh38
- UniGene ID
- Hs.592182
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001635.3 |
NP_001626.1
|
2, 3 |
370 |
|
NM_139316.2 |
NP_647477.1
|
2, 3 |
370 |
|
XM_006715689.3 |
XP_006715752.1
|
2, 3 |
317 |
|
XM_006715690.3 |
XP_006715753.1
|
2, 3 |
317 |
|
XM_006715691.3 |
XP_006715754.1
|
2, 3 |
317 |
|
XM_011515271.2 |
XP_011513573.1
|
2, 3 |
317 |
|
XM_017011995.1 |
XP_016867484.1
|
2, 3 |
317 |
|
XM_017011996.1 |
XP_016867485.1
|
2, 3 |
317 |
|
GenBank mRNA |
AF034996.1 |
- |
2, 3 |
223 |
|
AF498097.1 |
- |
2, 3 |
154 |
|
AK291447.1 |
- |
2, 3 |
174 |
|
BC034376.1 |
- |
2, 3 |
161 |
5181100 |
DA125756.1 |
- |
2, 3 |
370 |
|
U07616.1 |
- |
2, 3 |
264 |
|
X81438.1 |
- |
2, 3 |
228 |
|
View Full Product Details