Gene Transcripts

Gene Symbol
APOC4
Entrez Gene ID
346
Gene Name
apolipoprotein C4
Gene Aliases
APO-CIV, APOC-IV
Chromosome Location
Chr. 19: 44942238 - 44945496 on Build GRCh38
UniGene ID
Hs.731599
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001646.2 NP_001637.1 3 345
GenBank mRNA AV690709.1 - 3 294
BC020723.1 - 3 309 4700462
DB185657.1 - 3 345
EF371378.1 - 3 305

Gene Symbol
APOC4-APOC2
Entrez Gene ID
100533990
Gene Name
APOC4-APOC2 readthrough (NMD candidate)
Gene Aliases
Chromosome Location
Chr. 19: 44942238 - 44949565 on Build GRCh38
UniGene ID
Hs.731599
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_037932.1 3 345
GenBank mRNA AK126257.1 - 3 345

View Full Product Details