Gene Transcripts

Gene Symbol
AQP6
Entrez Gene ID
363
Gene Name
aquaporin 6
Gene Aliases
AQP2L, KID
Chromosome Location
Chr. 12: 49972837 - 49977139 on Build GRCh38
UniGene ID
Hs.54505
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001652.3 NP_001643.2 1 381
GenBank mRNA AL137716.1 - 4 2057
U48408.1 - 1 386

View Full Product Details