Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001172415.1 | NP_001165886.1 | 5 | 935 | |
NM_004323.5 | NP_004314.5 | 5 | 935 | ||
GenBank mRNA | AF022224.1 | - | 5 | 893 | |
AF116273.1 | - | 5 | 863 | ||
AK093238.1 | - | 2 | 1594 | ||
AK096038.1 | - | 6 | 698 | ||
AK222749.1 | - | 5 | 882 | ||
AK316577.1 | - | 5 | 861 | ||
BC001936.1 | - | 5 | 896 | 2823774 | |
BC014774.1 | - | 5 | 745 | ||
KU177932.1 | - | 5 | 635 | ||
KU177933.1 | - | 6 | 777 | ||
U46917.1 | - | 5 | 895 | ||
Z35491.1 | - | 5 | 913 |