Gene Transcripts

Gene Symbol
BNIP3L
Entrez Gene ID
665
Gene Name
BCL2 interacting protein 3 like
Gene Aliases
BNIP3a, NIX
Chromosome Location
Chr. 8: 26383007 - 26413128 on Build GRCh38
UniGene ID
Hs.131226
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004331.2 NP_004322.1 2 332
XM_005273617.3 XP_005273674.1 2 236
GenBank mRNA AB004788.1 - 2 253
AF060922.1 - 2 286
AF067396.1 - 2 290
AF079221.1 - 2 290
AF452712.2 - 2 305
AF536326.1 - 2 268
AK290213.1 - 2 248
AK300286.1 - 2 257
AK300727.1 - 2 271
AK315870.1 - 2 209
AK316315.1 - 2 236
AL132665.1 - 2 306
BC001559.1 - 2 232 3459327
BC009603.1 - 2 300 4134765
BT019500.1 - 2 208
BT019501.1 - 2 208
CD558918.1 - 2 441
CR456701.1 - 2 208

View Full Product Details