Gene Transcripts

Gene Symbol
CDH4
Entrez Gene ID
1002
Gene Name
cadherin 4
Gene Aliases
CAD4, R-CAD, RCAD
Chromosome Location
Chr. 20: 61252426 - 61940617 on Build GRCh38
UniGene ID
Hs.473231
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001252338.2 NP_001239267.1 6 881
NM_001252339.2 NP_001239268.1 6 1038
NM_001794.4 NP_001785.2 7 1066
GenBank mRNA AK091496.1 - 7 1262
AK124724.1 - 6 1021
AK308345.1 - 6 880
AK315192.1 - 7 1020
BC101651.1 - 7 1066 8069157
BC112150.1 - 7 1066 8327618
L34059.1 - 7 992

View Full Product Details