Gene Transcripts

Gene Symbol
HBEGF
Entrez Gene ID
1839
Gene Name
heparin binding EGF like growth factor
Gene Aliases
DTR, DTS, DTSF, HEGFL
Chromosome Location
Chr. 5: 140332843 - 140346603 on Build GRCh38
UniGene ID
Hs.592942
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001945.2 NP_001936.1 3 599
GenBank mRNA AK222688.1 - 3 599
AK307310.1 - 2 320
AK313202.1 - 3 600
BC033097.1 - 3 586 5424240
M60278.1 - 3 585

View Full Product Details