Gene Transcripts

Gene Symbol
LCN2
Entrez Gene ID
3934
Gene Name
lipocalin 2
Gene Aliases
24p3, MSFI, NGAL, p25
Chromosome Location
Chr. 9: 128149430 - 128153455 on Build GRCh38
UniGene ID
Hs.204238
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005564.4 NP_005555.2 5 588
GenBank mRNA AK301694.1 - 5 565
AK316217.1 - 5 565
AW778875.1 - 3 256
BC033089.1 - 5 552 5421124
BM977724.1 - 3 272
BX644845.1 - 5 601
CA454137.1 - 5 550
CR542092.1 - 5 492
EU644752.1 - 5 498
S75256.1 - 5 432
X83006.1 - 5 555

View Full Product Details