Gene Transcripts
- Gene Symbol
- MYBPC3
- Entrez Gene ID
- 4607
- Gene Name
- myosin binding protein C, cardiac
- Gene Aliases
- CMD1MM, CMH4, FHC, LVNC10, MYBP-C
- Chromosome Location
- Chr. 11: 47331406 - 47352702 on Build GRCh38
- UniGene ID
- Hs.524906
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000256.3 |
NP_000247.2
|
19, 20 |
1962 |
|
GenBank mRNA |
AK291569.1 |
- |
19, 20 |
1962 |
|
AK297843.1 |
- |
19, 20 |
1962 |
|
BC136543.1 |
- |
19, 20 |
1939 |
9020533 |
BC136546.1 |
- |
19, 20 |
1939 |
9020536 |
BC142685.1 |
- |
19, 20 |
1942 |
40148564 |
BC151211.1 |
- |
19, 20 |
1945 |
40147517 |
EF560722.1 |
- |
19, 20 |
1963 |
|
EU747718.1 |
- |
19, 20 |
1962 |
|
EU747719.1 |
- |
19, 20 |
1962 |
|
View Full Product Details