Gene Transcripts

Gene Symbol
NIT1
Entrez Gene ID
4817
Gene Name
nitrilase 1
Gene Aliases
Chromosome Location
Chr. 1: 161118072 - 161125446 on Build GRCh38
UniGene ID
Hs.146406
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001185092.1 NP_001172021.1 4 514
NM_001185093.1 NP_001172022.1 3 746
NM_001185094.1 NP_001172023.1 4 515
NM_005600.2 NP_005591.1 4 514
XM_005245214.2 XP_005245271.1 4 865
XM_005245215.4 XP_005245272.1 4 526
XM_005245216.3 XP_005245273.1 4 537
XM_006711341.3 XP_006711404.1 4 525
XM_017001371.1 XP_016856860.1 4 541
GenBank mRNA AF069987.1 - 4 485
AK296606.1 - 3 710
AK313323.1 - 4 481
BC046149.1 - 4 468 5764281
BI752623.1 - 4 515
BX416745.2 - 4 481
CB111935.1 - 4 514
CR541814.1 - 3 409
CR541846.1 - 3 409

View Full Product Details