Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005859.4 | NP_005850.1 | 1 | 731 | |
GenBank mRNA | BC036087.1 | - | 1 | 684 | |
BC110574.1 | - | 2 | 561 | ||
BC110594.1 | - | 2 | 610 | ||
BT019388.1 | - | 1 | 672 | ||
M96684.1 | - | 1 | 731 |