Gene Transcripts

Gene Symbol
TACR1
Entrez Gene ID
6869
Gene Name
tachykinin receptor 1
Gene Aliases
NK1R, NKIR, SPR, TAC1R
Chromosome Location
Chr. 2: 75046463 - 75199519 on Build GRCh38
UniGene ID
Hs.633301
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001058.3 NP_001049.1 4 1417
NM_015727.2 NP_056542.1 4 1417
GenBank mRNA AK289765.1 - 4 1386
AY462098.1 - 4 832
BC074911.2 - 4 854 30915523
BC074912.2 - 4 854 30915310
BC095528.1 - 4 829
M74290.1 - 4 1042
M76675.1 - 4 832
M81797.1 - 4 876
M84425.1 - 4 954
M84426.1 - 4 954
S62045.1 - 4 896

View Full Product Details