Gene Transcripts

Gene Symbol
ZNF22
Entrez Gene ID
7570
Gene Name
zinc finger protein 22
Gene Aliases
HKR-T1, KOX15, ZNF422, Zfp422
Chromosome Location
Chr. 10: 45000825 - 45005329 on Build GRCh38
UniGene ID
Hs.462693
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006963.4 NP_008894.2 2 387
GenBank mRNA AK291508.1 - 2 289
AK312278.1 - 2 289
BC010642.1 - 2 277 3852749
BC026193.1 - 7 1183
BC041139.1 - 2 738 5751460
BC053687.1 - 2 281 5214077
BT009806.1 - 1 144
S50223.1 - 1 233

View Full Product Details