Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_021969.2 | NP_068804.1 | 1 | 633 | |
XM_011542297.2 | XP_011540599.1 | 1 | 633 | ||
GenBank mRNA | AK310048.1 | - | 1 | 634 | |
BC030207.1 | - | 1 | 523 | 5222402 | |
HQ692833.1 | - | 1 | 497 |