Gene Transcripts

Gene Symbol
ASAP2
Entrez Gene ID
8853
Gene Name
ArfGAP with SH3 domain, ankyrin repeat and PH domain 2
Gene Aliases
AMAP2, CENTB3, DDEF2, PAG3, PAP, Pap-alpha, SHAG1
Chromosome Location
Chr. 2: 9206765 - 9405683 on Build GRCh38
UniGene ID
Hs.555902
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135191.1 NP_001128663.1 26 3051
NM_003887.2 NP_003878.1 27 3186
XM_006711898.1 XP_006711961.1 28 2948
XM_006711901.3 XP_006711964.1 28 2798
XM_006711902.1 XP_006711965.1 27 2813
XM_011510403.2 XP_011508705.1 28 2969
XM_011510404.2 XP_011508706.1 27 2959
XM_011510405.2 XP_011508707.1 27 2895
XM_011510406.2 XP_011508708.1 29 2902
XM_011510407.2 XP_011508709.1 27 2834
XM_011510409.1 XP_011508711.1 28 2840
GenBank mRNA AB007860.1 - 27 3186
AK091121.1 - 5 597
AK124900.1 - 3 1344
BC063308.1 - 26 3004 30345039

View Full Product Details