Gene Transcripts
- Gene Symbol
- GYG2
- Entrez Gene ID
- 8908
- Gene Name
- glycogenin 2
- Gene Aliases
- GN-2, GN2
- Chromosome Location
- Chr. X: 2828712 - 2882820 on Build GRCh38
- UniGene ID
- Hs.567381
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001079855.1 |
NP_001073324.1
|
6, 7 |
884 |
|
NM_001184702.1 |
NP_001171631.1
|
6, 7 |
884 |
|
NM_001184703.1 |
NP_001171632.1
|
7, 8 |
977 |
|
NM_001184704.1 |
NP_001171633.1
|
5, 6 |
742 |
|
NM_003918.2 |
NP_003909.2
|
7, 8 |
977 |
|
XM_011545600.2 |
XP_011543902.1
|
6, 7 |
815 |
|
XM_017029927.1 |
XP_016885416.1
|
7, 8 |
927 |
|
XM_017029928.1 |
XP_016885417.1
|
7, 8 |
902 |
|
XM_017029929.1 |
XP_016885418.1
|
7, 8 |
1087 |
|
XM_017029930.1 |
XP_016885419.1
|
6, 7 |
792 |
|
XM_017029931.1 |
XP_016885420.1
|
6, 7 |
815 |
|
GenBank mRNA |
AK098046.1 |
- |
5, 6 |
650 |
|
AK292496.1 |
- |
6, 7 |
792 |
|
BC023152.1 |
- |
6, 7 |
751 |
3923041 |
DQ424901.1 |
- |
5, 6 |
695 |
|
U94358.1 |
- |
3, 4 |
373 |
|
U94360.1 |
- |
3, 4 |
432 |
|
U94361.1 |
- |
2, 3 |
150 |
|
U94362.1 |
- |
7, 8 |
978 |
|
U94363.1 |
- |
6, 7 |
758 |
|
View Full Product Details