Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_199328.2 | NP_955360.1 | 1 | 528 | |
GenBank mRNA | AK022269.1 | - | 1 | 451 | |
AK223512.1 | - | 1 | 422 | ||
AY358707.1 | - | 1 | 398 | ||
BC020866.1 | - | 1 | 411 | 4594155 | |
BC058004.1 | - | 1 | 377 | 4606880 | |
DA629490.1 | - | 1 | 528 | ||
GQ472207.1 | - | 1 | 411 |