Gene Transcripts
- Gene Symbol
- ADIPOQ
- Entrez Gene ID
- 9370
- Gene Name
- adiponectin, C1Q and collagen domain containing
- Gene Aliases
- ACDC, ACRP30, ADIPQTL1, ADPN, APM-1, APM1, GBP28
- Chromosome Location
- Chr. 3: 186842674 - 186858463 on Build GRCh38
- UniGene ID
- Hs.80485
- Species
- Homo sapiens
- Gene Symbol
- ADIPOQ-AS1
- Entrez Gene ID
- 100874095
- Gene Name
- ADIPOQ antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 3: 186851886 - 186856123 on Build GRCh38
- UniGene ID
- Hs.638968
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046662.1 |
|
1 |
1718 |
|
GenBank mRNA |
BC036509.1 |
- |
2 |
1854 |
|
View Full Product Details