Gene Transcripts

Gene Symbol
TM9SF2
Entrez Gene ID
9375
Gene Name
transmembrane 9 superfamily member 2
Gene Aliases
P76
Chromosome Location
Chr. 13: 99501374 - 99564048 on Build GRCh38
UniGene ID
Hs.607896
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004800.2 NP_004791.1 6 921
GenBank mRNA AK093507.1 - 6 518
AK290514.1 - 6 876
AK315210.1 - 6 876
BC110656.1 - 6 878 3855486
BC130293.1 - 6 755 40146324
U81006.1 - 6 821

View Full Product Details