Gene Transcripts

Gene Symbol
CHST2
Entrez Gene ID
9435
Gene Name
carbohydrate sulfotransferase 2
Gene Aliases
C6ST, GST-2, GST2, Gn6ST-1, HEL-S-75, glcNAc6ST-1
Chromosome Location
Chr. 3: 143119776 - 143124014 on Build GRCh38
UniGene ID
Hs.529249
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004267.4 NP_004258.2 2 2350
GenBank mRNA AB014679.2 - 2 1659
AB014680.2 - 2 1659
AB021124.1 - 2 2795
AF083066.1 - 1 1468
AK023268.1 - 2 2300
BC017499.1 - 2 2031
BC036930.1 - 2 2261
BC068604.1 - 2 2300
BC105010.1 - 1 1430 8144013
BC105012.1 - 1 1430 8144015
EU668328.1 - 1 1430

View Full Product Details