Gene Transcripts
- Gene Symbol
- RBM8A
- Entrez Gene ID
- 9939
- Gene Name
- RNA binding motif protein 8A
- Gene Aliases
- BOV-1A, BOV-1B, BOV-1C, C1DELq21.1, DEL1q21.1, MDS014, RBM8, RBM8B, TAR, Y14, ZNRP, ZRNP1
- Chromosome Location
- Chr. 1: 145921556 - 145927536 on Build GRCh38
- UniGene ID
- Hs.591455
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_005105.4 |
NP_005096.1
|
3, 4 |
322 |
|
GenBank mRNA |
AF127761.1 |
- |
3, 4 |
224 |
|
AF161463.1 |
- |
3, 4 |
236 |
|
AF182415.1 |
- |
3, 4 |
241 |
|
AF198620.1 |
- |
3, 4 |
243 |
|
AF231511.1 |
- |
3, 4 |
229 |
|
AF299118.1 |
- |
3, 4 |
212 |
|
AK075009.1 |
- |
3, 4 |
243 |
|
BC017088.2 |
- |
3, 4 |
284 |
3850355 |
BC071577.1 |
- |
3, 4 |
270 |
|
CR541805.1 |
- |
3, 4 |
212 |
|
CR541823.1 |
- |
3, 4 |
212 |
|
DB286556.1 |
- |
3, 4 |
322 |
|
KF051020.1 |
- |
3, 4 |
212 |
|
View Full Product Details