Gene Transcripts

Gene Symbol
RBM8A
Entrez Gene ID
9939
Gene Name
RNA binding motif protein 8A
Gene Aliases
BOV-1A, BOV-1B, BOV-1C, C1DELq21.1, DEL1q21.1, MDS014, RBM8, RBM8B, TAR, Y14, ZNRP, ZRNP1
Chromosome Location
Chr. 1: 145921556 - 145927536 on Build GRCh38
UniGene ID
Hs.591455
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005105.4 NP_005096.1 4, 5 463
GenBank mRNA AF127761.1 - 4, 5 365
AF161463.1 - 4, 5 377
AF182415.1 - 4, 5 382
AF198620.1 - 4, 5 384
AF231511.1 - 4, 5 370
AF299118.1 - 4, 5 353
AK075009.1 - 4, 5 384
BC017088.2 - 4, 5 425 3850355
BC017770.1 - 2, 3 703
BC071577.1 - 4, 5 411
CR541805.1 - 4, 5 353
CR541823.1 - 4, 5 353
DB286556.1 - 4, 5 463
KF051020.1 - 4, 5 353

View Full Product Details