Gene Transcripts

Gene Symbol
PLIN3
Entrez Gene ID
10226
Gene Name
perilipin 3
Gene Aliases
M6PRBP1, PP17, TIP47
Chromosome Location
Chr. 19: 4838334 - 4867768 on Build GRCh38
UniGene ID
Hs.140452
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164189.1 NP_001157661.1 7 1083
NM_001164194.1 NP_001157666.1 7 1047
NM_005817.4 NP_005808.3 7 1083
GenBank mRNA AF051314.1 - 3 395
AF051315.1 - 3 371
AF055574.1 - 7 959
AF057140.1 - 7 980
AK223054.1 - 7 1013
AK225045.1 - 7 949
AK225400.1 - 7 964
AK226088.1 - 7 961
AK291104.1 - 7 1040
AK312740.1 - 7 1009
BC001590.2 - 7 956 2987965
BC005818.2 - 7 972 3833411
BC007566.1 - 7 972 3028104
BC019278.1 - 7 972 2905275
BT007235.1 - 6 906

View Full Product Details