Gene Transcripts
- Gene Symbol
- AGR2
- Entrez Gene ID
- 10551
- Gene Name
- anterior gradient 2, protein disulphide isomerase family member
- Gene Aliases
- AG2, GOB-4, HAG-2, HEL-S-116, PDIA17, XAG-2
- Chromosome Location
- Chr. 7: 16791811 - 16805114 on Build GRCh38
- UniGene ID
- Hs.530009
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_006408.3 |
NP_006399.1
|
4, 5 |
440 |
|
XM_005249581.4 |
XP_005249638.1
|
4, 5 |
409 |
|
GenBank mRNA |
AF007791.1 |
- |
3, 4 |
263 |
|
AF038451.1 |
- |
4, 5 |
311 |
|
AF087879.1 |
- |
4, 5 |
402 |
|
AF088867.1 |
- |
4, 5 |
400 |
|
AF115926.1 |
- |
4, 5 |
310 |
|
AK223067.1 |
- |
4, 5 |
331 |
|
AK312942.1 |
- |
4, 5 |
321 |
|
AY359009.1 |
- |
4, 5 |
400 |
|
BC015503.1 |
- |
4, 5 |
309 |
3852448 |
BT007048.1 |
- |
3, 4 |
253 |
|
FJ224307.1 |
- |
3, 4 |
263 |
|
KJ767789.1 |
- |
4, 5 |
414 |
|
View Full Product Details