Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001190996.1 | NP_001177925.1 | 9 | 1171 | |
NM_006409.3 | NP_006400.2 | 9 | 1173 | ||
GenBank mRNA | AB209059.1 | - | 9 | 1106 | |
AK223015.1 | - | 8 | 997 | ||
BC039594.1 | - | 9 | 1136 | 6018804 | |
BC047889.1 | - | 9 | 1106 | 6473050 | |
BC054027.1 | - | 9 | 1105 | 6514889 | |
EU668326.1 | - | 9 | 1159 | ||
GQ472234.1 | - | 8 | 997 | ||
Y08999.1 | - | 8 | 1045 |