Gene Transcripts

Gene Symbol
IFI44
Entrez Gene ID
10561
Gene Name
interferon induced protein 44
Gene Aliases
MTAP44, TLDC5, p44
Chromosome Location
Chr. 1: 78648554 - 78664078 on Build GRCh38
UniGene ID
Hs.82316
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006417.4 NP_006408.3 6 1087
NR_135640.1 5 623
NR_135641.1 6 1090
XM_005270380.3 XP_005270437.1 6 2325
XM_011540516.2 XP_011538818.1 6 2325
XM_017000083.1 XP_016855572.1 6 2325
GenBank mRNA AK302527.1 - 6 1048
AK316278.1 - 5 580
AK316476.1 - 6 1045
BC022870.1 - 6 1052 4071404
CA426666.1 - 4 652

View Full Product Details