Gene Transcripts
- Gene Symbol
- FARS2
- Entrez Gene ID
- 10667
- Gene Name
- phenylalanyl-tRNA synthetase 2, mitochondrial
- Gene Aliases
- COXPD14, FARS1, HSPC320, PheRS
- Chromosome Location
- Chr. 6: 5261001 - 5771592 on Build GRCh38
- UniGene ID
- Hs.484547
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001318872.1 |
NP_001305801.1
|
5, 6 |
1425 |
|
NM_006567.4 |
NP_006558.1
|
5, 6 |
1419 |
|
XM_005248812.3 |
XP_005248869.1
|
6, 7 |
1483 |
|
XM_011514247.2 |
XP_011512549.1
|
5, 6 |
1394 |
|
XM_011514248.2 |
XP_011512550.1
|
5, 6 |
1394 |
|
XM_011514249.1 |
XP_011512551.1
|
5, 6 |
1394 |
|
XM_017010186.1 |
XP_016865675.1
|
5, 6 |
2202 |
|
XM_017010187.1 |
XP_016865676.1
|
6, 7 |
2204 |
|
GenBank mRNA |
AF097441.1 |
- |
4, 5 |
1075 |
|
AF161438.1 |
- |
4, 5 |
744 |
|
AI338405.1 |
- |
2, 3 |
448 |
|
AK223423.1 |
- |
5, 6 |
1219 |
|
AK312454.1 |
- |
5, 6 |
1399 |
|
BC020239.1 |
- |
5, 6 |
1219 |
5258249 |
BC021112.1 |
- |
5, 6 |
1219 |
5088776 |
CA432122.1 |
- |
2, 3 |
470 |
|
CR542279.1 |
- |
4, 5 |
1063 |
|
View Full Product Details