Gene Transcripts

Gene Symbol
PDIA5
Entrez Gene ID
10954
Gene Name
protein disulfide isomerase family A member 5
Gene Aliases
PDIR
Chromosome Location
Chr. 3: 123067009 - 123162106 on Build GRCh38
UniGene ID
Hs.477352
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006810.3 NP_006801.1 16 1586
NR_028444.1 15 1430
GenBank mRNA AK054963.1 - 5 1534
BC001625.1 - 15 1377 3537659
CA414869.1 - 2 288
CB158962.1 - 2 284
D49490.1 - 16 1486

View Full Product Details