Gene Transcripts

Gene Symbol
LSM6
Entrez Gene ID
11157
Gene Name
LSM6 homolog, U6 small nuclear RNA and mRNA degradation associated
Gene Aliases
YDR378C
Chromosome Location
Chr. 4: 146175683 - 146204436 on Build GRCh38
UniGene ID
Hs.190520
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007080.2 NP_009011.1 2 193
XM_017007672.1 XP_016863161.1 2 168
XM_017007673.1 XP_016863162.1 2 493
GenBank mRNA AF182292.1 - 2 135
AJ238098.1 - 2 135
AK312126.1 - 2 128
BC016026.1 - 2 147 3910698
BC019894.1 - 2 773
DB465201.1 - 2 194

View Full Product Details