Gene Transcripts

Gene Symbol
WDHD1
Entrez Gene ID
11169
Gene Name
WD repeat and HMG-box DNA binding protein 1
Gene Aliases
AND-1, AND1, CHTF4, CTF4
Chromosome Location
Chr. 14: 54938938 - 55027105 on Build GRCh38
UniGene ID
Hs.385998
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008396.2 NP_001008397.1 5 479
NM_007086.3 NP_009017.1 6 572
XM_006720012.2 XP_006720075.1 6 570
XM_011536373.2 XP_011534675.1 5 483
XM_017020940.1 XP_016876429.1 5 483
XM_017020941.1 XP_016876430.1 3 293
GenBank mRNA AJ006266.1 - 5 521
AK293005.1 - 6 539
BC000622.2 - 6 537
BC005040.1 - 6 537
BC043349.1 - 6 560
BC063041.1 - 6 548 6514641
DA189308.1 - 5 479

View Full Product Details