Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001303050.1 | NP_001289979.1 | 4 | 660 | |
NM_012326.3 | NP_036458.2 | 4 | 861 | ||
GenBank mRNA | AB025186.1 | - | 4 | 575 | |
AK312342.1 | - | 4 | 595 | ||
BC011557.1 | - | 4 | 560 | 4561018 | |
CR536523.1 | - | 3 | 422 | ||
CR541845.1 | - | 3 | 422 | ||
Y11174.1 | - | 4 | 446 |