Gene Transcripts

Gene Symbol
TNIK
Entrez Gene ID
23043
Gene Name
TRAF2 and NCK interacting kinase
Gene Aliases
Chromosome Location
Chr. 3: 171058414 - 171460408 on Build GRCh38
UniGene ID
Hs.34024
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161560.2 NP_001155032.1 29 3959
NM_001161561.2 NP_001155033.1 29 3896
NM_001161562.2 NP_001155034.1 28 3872
NM_001161563.2 NP_001155035.1 29 3818
NM_001161564.2 NP_001155036.1 28 3794
NM_001161565.2 NP_001155037.1 28 3731
NM_001161566.2 NP_001155038.1 27 3707
NM_015028.3 NP_055843.1 30 3983
GenBank mRNA AB011123.2 - 30 3904
AF172264.1 - 30 3638
AF172265.1 - 29 3551
AF172266.1 - 29 3473
AF172267.1 - 29 3614
AF172268.1 - 28 3386
AF172269.1 - 28 3527
AF172270.1 - 28 3449
AF172271.1 - 27 3362
AK026470.1 - 4 757
AK291056.1 - 28 3731
BC019081.2 - 9 1102
BC055427.1 - 9 1671
BC150256.1 - 30 3904 9007184

View Full Product Details