Gene Transcripts
- Gene Symbol
- CIC
- Entrez Gene ID
- 23152
- Gene Name
- capicua transcriptional repressor
- Gene Aliases
- Chromosome Location
- Chr. 19: 42268537 - 42295796 on Build GRCh38
- UniGene ID
- Hs.388236
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001304815.1 |
NP_001291744.1
|
10, 11 |
4265 |
|
NM_015125.4 |
NP_055940.3
|
9, 10 |
1510 |
|
XM_005258673.2 |
XP_005258730.1
|
9, 10 |
1602 |
|
XM_005258674.2 |
XP_005258731.1
|
9, 10 |
1602 |
|
XM_005258675.2 |
XP_005258732.1
|
9, 10 |
1602 |
|
XM_011526660.2 |
XP_011524962.1
|
10, 11 |
4307 |
|
XM_011526661.2 |
XP_011524963.1
|
10, 11 |
4307 |
|
XM_011526662.2 |
XP_011524964.1
|
10, 11 |
4307 |
|
XM_011526663.2 |
XP_011524965.1
|
10, 11 |
4307 |
|
XM_011526664.2 |
XP_011524966.1
|
10, 11 |
4307 |
|
XM_011526665.2 |
XP_011524967.1
|
10, 11 |
4307 |
|
XM_011526666.2 |
XP_011524968.1
|
10, 11 |
4307 |
|
XM_011526667.2 |
XP_011524969.1
|
10, 11 |
4307 |
|
GenBank mRNA |
AB002304.1 |
- |
6, 7 |
1001 |
|
AF363689.1 |
- |
9, 10 |
1510 |
|
View Full Product Details