Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_015340.3 | NP_056155.1 | 15 | 1873 | |
XM_005265006.1 | XP_005265063.1 | 14 | 1829 | ||
XM_011533554.2 | XP_011531856.1 | 15 | 1892 | ||
XM_017006042.1 | XP_016861531.1 | 15 | 1892 | ||
GenBank mRNA | AK025465.1 | - | 9 | 1173 | |
AK026113.1 | - | 2 | 96 | ||
AK308146.1 | - | 13 | 1697 | ||
BC025989.2 | - | 15 | 1880 | 4822546 | |
D21851.1 | - | 15 | 1873 |