Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001301140.1 | NP_001288069.1 | 3 | 1418 | |
NM_001301141.1 | NP_001288070.1 | 3 | 1418 | ||
NM_001320042.1 | NP_001306971.1 | 3 | 1418 | ||
NM_014301.4 | NP_055116.1 | 4 | 1514 | ||
NM_213595.3 | NP_998760.1 | 3 | 1418 | ||
NR_135127.1 | 4 | 1514 | |||
XM_011538100.2 | XP_011536402.1 | 4 | 1514 | ||
XM_011538101.2 | XP_011536403.1 | 3 | 1418 | ||
GenBank mRNA | AK000574.1 | - | 3 | 261 | |
AK022758.1 | - | 2 | 1842 | ||
AK057251.1 | - | 3 | 1418 | ||
AK293202.1 | - | 3 | 271 | ||
AK297862.1 | - | 3 | 290 | ||
AY009127.1 | - | 4 | 371 | ||
AY009128.1 | - | 3 | 275 | ||
BC011906.2 | - | 3 | 274 | 4136262 | |
BC061903.1 | - | 3 | 267 | 4183674 | |
BG702203.1 | - | 3 | 269 | ||
BI597126.1 | - | 3 | 369 | ||
BM423850.1 | - | 3 | 264 | ||
BM921073.1 | - | 3 | 359 | ||
BP378051.1 | - | 3 | 290 | ||
BU602914.1 | - | 3 | 279 | ||
DB095495.1 | - | 3 | 285 | ||
DB493673.1 | - | 3 | 348 | ||
EU329002.1 | - | 4 | 371 | ||
U47101.1 | - | 2 | 121 |