Gene Transcripts

Gene Symbol
HEBP2
Entrez Gene ID
23593
Gene Name
heme binding protein 2
Gene Aliases
C6ORF34B, C6orf34, PP23, SOUL
Chromosome Location
Chr. 6: 138404199 - 138415593 on Build GRCh38
UniGene ID
Hs.486589
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001326380.1 NP_001313309.1 2 261
NM_001326381.1 NP_001313310.1 2 492
NM_014320.2 NP_055135.1 2 492
XM_017010631.1 XP_016866120.1 2 465
GenBank mRNA AF117616.1 - 2 470
AF411610.1 - 2 132
AK057928.1 - 2 407
AY427823.1 - 2 374
BC008205.1 - 2 356 3869026
BC010290.1 - 2 369 3445763
BC016806.1 - 2 472
BC016807.1 - 2 472
BC093037.1 - 2 355 30333215
BF974311.1 - 492

View Full Product Details