Gene Transcripts
- Gene Symbol
- FBXL2
- Entrez Gene ID
- 25827
- Gene Name
- F-box and leucine rich repeat protein 2
- Gene Aliases
- FBL2, FBL3
- Chromosome Location
- Chr. 3: 33277422 - 33422371 on Build GRCh38
- UniGene ID
- Hs.475872
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001171713.1 |
NP_001165184.1
|
5, 6 |
354 |
|
NM_012157.3 |
NP_036289.3
|
5, 6 |
357 |
|
XM_005265014.2 |
XP_005265071.1
|
5, 6 |
377 |
|
XM_005265015.3 |
XP_005265072.1
|
5, 6 |
578 |
|
XM_005265016.2 |
XP_005265073.1
|
5, 6 |
322 |
|
XM_005265017.3 |
XP_005265074.1
|
5, 6 |
354 |
|
XM_011533559.2 |
XP_011531861.1
|
6, 7 |
523 |
|
XM_017006092.1 |
XP_016861581.1
|
6, 7 |
415 |
|
XM_017006093.1 |
XP_016861582.1
|
5, 6 |
405 |
|
XM_017006094.1 |
XP_016861583.1
|
6, 7 |
438 |
|
GenBank mRNA |
AF174589.1 |
- |
6, 7 |
328 |
|
AF176518.1 |
- |
5, 6 |
292 |
|
AF186273.1 |
- |
5, 6 |
307 |
|
AK001271.1 |
- |
5, 6 |
357 |
|
AK001438.1 |
- |
5, 6 |
325 |
|
AK297534.1 |
- |
5, 6 |
322 |
|
AK298967.1 |
- |
5, 6 |
323 |
|
AK299665.1 |
- |
6, 7 |
440 |
|
AK302218.1 |
- |
7, 8 |
585 |
|
AK316229.1 |
- |
5, 6 |
378 |
|
AK316383.1 |
- |
6, 7 |
497 |
|
AU124816.1 |
- |
5, 6 |
357 |
|
BC031556.1 |
- |
5, 6 |
301 |
5197151 |
CR457121.1 |
- |
5, 6 |
286 |
|
View Full Product Details