Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_015490.3 | NP_056305.1 | 13 | 1662 | |
GenBank mRNA | AF274863.1 | - | 13 | 1658 | |
AK128343.1 | - | 7 | 1796 | ||
AL080141.1 | - | 6 | 770 | ||
BC034946.1 | - | 13 | 2218 | ||
BC044569.1 | - | 13 | 2211 | ||
BC053949.1 | - | 13 | 2211 | ||
BC071580.1 | - | 13 | 2138 | ||
BC144313.1 | - | 12 | 1600 | 9052833 | |
BX640914.1 | - | 13 | 1559 |