Gene Transcripts

Gene Symbol
FBXO24
Entrez Gene ID
26261
Gene Name
F-box protein 24
Gene Aliases
FBX24
Chromosome Location
Chr. 7: 100582490 - 100601117 on Build GRCh38
UniGene ID
Hs.283764
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163499.1 NP_001156971.1 5, 6 832
NM_012172.4 NP_036304.2 5, 6 982
NM_033506.2 NP_277041.1 5, 6 1093
XM_005250259.3 XP_005250316.1 5, 6 1267
XM_011516022.1 XP_011514324.1 5, 6 825
XM_011516023.2 XP_011514325.1 4, 5 1084
XM_017011961.1 XP_016867450.1 6, 7 2568
GenBank mRNA AF174604.1 - 5, 6 758
AK292240.1 - 5, 6 1100
AK301864.1 - 5, 6 926
AK302252.1 - 5, 6 832
AL136811.1 - 5, 6 1038
BC067293.1 - 5, 6 1033 30383439

View Full Product Details