Gene Transcripts

Gene Symbol
NBEA
Entrez Gene ID
26960
Gene Name
neurobeachin
Gene Aliases
BCL8B, LYST2
Chromosome Location
Chr. 13: 34942287 - 35672737 on Build GRCh38
UniGene ID
Hs.491172
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015678.4 NP_056493.3 22 3958
XM_005266346.4 XP_005266403.1 22 3958
XM_005266347.4 XP_005266404.2 22 3958
XM_005266348.4 XP_005266405.2 22 3958
XM_006719803.3 XP_006719866.1 22 3958
XM_006719805.3 XP_006719868.1 22 3958
XM_006719806.3 XP_006719869.1 22 3958
XM_011535046.2 XP_011533348.1 14 2089
XM_011535047.2 XP_011533349.1 22 3958
XM_017020544.1 XP_016876033.1 22 3958
XM_017020545.1 XP_016876034.1 22 3958
XM_017020546.1 XP_016876035.1 22 3246
GenBank mRNA AF467288.1 - 23 3630

View Full Product Details