Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013245.2 | NP_037377.1 | 11 | 1400 | |
GenBank mRNA | AF112215.1 | - | 11 | 1355 | |
AF132747.1 | - | 11 | 1408 | ||
AF155740.1 | - | 10 | 1258 | ||
AF159063.1 | - | 11 | 1400 | ||
AF255952.1 | - | 11 | 1400 | ||
AK000200.1 | - | 7 | 832 | ||
AK025850.1 | - | 6 | 704 | ||
AK315026.1 | - | 11 | 1428 | ||
AL133634.1 | - | 6 | 761 | ||
BC035121.1 | - | 2 | 1551 | ||
BC047932.1 | - | 11 | 1378 | 5753632 |