Gene Transcripts
- Gene Symbol
- LSM1
- Entrez Gene ID
- 27257
- Gene Name
- LSM1 homolog, mRNA degradation associated
- Gene Aliases
- CASM, YJL124C
- Chromosome Location
- Chr. 8: 38163321 - 38176730 on Build GRCh38
- UniGene ID
- Hs.425311
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_014462.2 |
NP_055277.1
|
2, 3 |
526 |
|
NR_045493.1 |
|
2, 3 |
248 |
|
GenBank mRNA |
AF000177.1 |
- |
2, 3 |
280 |
|
AF147348.1 |
- |
2, 3 |
169 |
|
AJ238094.1 |
- |
2, 3 |
304 |
|
AK312159.1 |
- |
2, 3 |
279 |
|
BC001767.1 |
- |
2, 3 |
213 |
3353833 |
BP376421.1 |
- |
2, 3 |
489 |
|
BU941962.1 |
- |
2, 3 |
251 |
|
KU178606.1 |
- |
2, 3 |
116 |
|
View Full Product Details