Gene Transcripts

Gene Symbol
DNAJC15
Entrez Gene ID
29103
Gene Name
DnaJ heat shock protein family (Hsp40) member C15
Gene Aliases
DNAJD1, HSD18, MCJ
Chromosome Location
Chr. 13: 43023226 - 43109170 on Build GRCh38
UniGene ID
Hs.438830
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013238.2 NP_037370.2 5 751
GenBank mRNA AF126743.1 - 6 773
AJ270691.1 - 2 117
AK311866.1 - 5 387
AY248898.1 - 5 751
AY512565.1 - 5 444
BC010910.1 - 5 480 4332619
BC095400.1 - 5 370 30530999

View Full Product Details