Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013320.2 | NP_037452.1 | 9 | 1359 | |
XM_017019241.1 | XP_016874730.1 | 8 | 1191 | ||
XM_017019242.1 | XP_016874731.1 | 9 | 1359 | ||
GenBank mRNA | AF117210.1 | - | 9 | 1268 | |
AK223342.1 | - | 9 | 1286 | ||
AK296523.1 | - | 2 | 141 | ||
AK313466.1 | - | 9 | 1289 | ||
BC033799.1 | - | 9 | 1261 | 5172046 |