Gene Transcripts
- Gene Symbol
- SH3KBP1
- Entrez Gene ID
- 30011
- Gene Name
- SH3 domain containing kinase binding protein 1
- Gene Aliases
- CD2BP3, CIN85, GIG10, HSB-1, HSB1, MIG18
- Chromosome Location
- Chr. X: 19533965 - 19887626 on Build GRCh38
- UniGene ID
- Hs.726365
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001024666.2 |
NP_001019837.1
|
13, 14 |
1493 |
|
NM_001184960.1 |
NP_001171889.1
|
9, 10 |
903 |
|
NM_031892.2 |
NP_114098.1
|
14, 15 |
1805 |
|
XM_005274494.2 |
XP_005274551.1
|
15, 16 |
1746 |
|
XM_011545498.2 |
XP_011543800.1
|
16, 17 |
1803 |
|
XM_011545499.2 |
XP_011543801.1
|
15, 16 |
1611 |
|
XM_011545502.2 |
XP_011543804.1
|
9, 10 |
909 |
|
XM_017029459.1 |
XP_016884948.1
|
15, 16 |
1543 |
|
XM_017029460.1 |
XP_016884949.1
|
15, 16 |
1586 |
|
XM_017029461.1 |
XP_016884950.1
|
14, 15 |
1529 |
|
XM_017029463.1 |
XP_016884952.1
|
13, 14 |
1513 |
|
XM_017029465.1 |
XP_016884954.1
|
14, 15 |
1772 |
|
XM_017029469.1 |
XP_016884958.1
|
9, 10 |
859 |
|
GenBank mRNA |
AF230904.1 |
- |
14, 15 |
1780 |
|
AF329267.1 |
- |
8, 9 |
706 |
|
AF329268.1 |
- |
8, 9 |
706 |
|
AF542051.1 |
- |
13, 14 |
1456 |
|
AK293234.1 |
- |
13, 14 |
1433 |
|
AK293237.1 |
- |
9, 10 |
803 |
|
AK300189.1 |
- |
9, 10 |
813 |
|
AK313308.1 |
- |
14, 15 |
1733 |
|
AY423734.1 |
- |
15, 16 |
1703 |
|
AY542305.1 |
- |
14, 15 |
1540 |
|
BC015806.1 |
- |
14, 15 |
1707 |
3906722 |
View Full Product Details