Gene Transcripts

Gene Symbol
TAS2R4
Entrez Gene ID
50832
Gene Name
taste 2 receptor member 4
Gene Aliases
T2R4
Chromosome Location
Chr. 7: 141778489 - 141779388 on Build GRCh38
UniGene ID
Hs.735690
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016944.1 NP_058640.1 1 478
GenBank mRNA BC130439.1 - 1 525
BC136508.1 - 1 525 9020497

View Full Product Details