Gene Transcripts

Gene Symbol
TRAPPC12
Entrez Gene ID
51112
Gene Name
trafficking protein particle complex 12
Gene Aliases
CGI-87, TTC-15, TTC15
Chromosome Location
Chr. 2: 3379245 - 3479571 on Build GRCh38
UniGene ID
Hs.252713
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321102.1 NP_001308031.1 2, 3 1216
NM_016030.5 NP_057114.5 2, 3 1253
XM_011510350.2 XP_011508652.1 2, 3 1683
XM_011510352.2 XP_011508654.1 2, 3 1683
XM_011510353.2 XP_011508655.1 2, 3 1683
XM_011510354.2 XP_011508656.1 2, 3 1683
XM_017004257.1 XP_016859746.1 2, 3 1683
GenBank mRNA AK094181.1 - 3, 4 1203
AK094242.1 - 1, 2 627
AK098327.1 - 2, 3 1218
BC014164.2 - 2, 3 1130 4548681
BC017475.1 - 1, 2 251

View Full Product Details