Gene Transcripts
- Gene Symbol
- HSD17B12
- Entrez Gene ID
- 51144
- Gene Name
- hydroxysteroid 17-beta dehydrogenase 12
- Gene Aliases
- KAR, SDR12C1
- Chromosome Location
- Chr. 11: 43680593 - 43856619 on Build GRCh38
- UniGene ID
- Hs.132513
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_016142.2 |
NP_057226.1
|
2, 3 |
444 |
|
XM_011520156.1 |
XP_011518458.1
|
2, 3 |
438 |
|
XM_017017881.1 |
XP_016873370.1
|
2, 3 |
124 |
|
GenBank mRNA |
AF078850.1 |
- |
2, 3 |
282 |
|
AK027882.1 |
- |
|
338 |
|
AK074952.1 |
- |
2, 3 |
296 |
|
AK075011.1 |
- |
2, 3 |
329 |
|
AK075216.1 |
- |
2, 3 |
305 |
|
AK222881.1 |
- |
2, 3 |
266 |
|
AK301663.1 |
- |
3, 4 |
235 |
|
BC012043.1 |
- |
2, 3 |
267 |
3622879 |
BC012536.1 |
- |
2, 3 |
260 |
|
View Full Product Details